Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108331.2 | NP_001101801.2 | 1 - 2 | 75 | 89 | |
XM_006249179.2 | XP_006249241.1 | 1 - 2 | 104 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |