Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109226.1 | NP_001102696.1 | 3 - 4 | 962 | 95 | |
XM_006236220.2 | XP_006236282.1 | 3 - 4 | 1169 | 95 | ||
XM_008762817.1 | XP_008761039.1 | 3 - 4 | 853 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |