Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024323.1 | NP_001019494.1 | 1 - 2 | 235 | 68 | |
GenBank mRNA | BC086566.1 | - | 1 - 2 | 235 | 7112589 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |