Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077668.2 | NP_001071136.1 | 2 - 3 | 326 | 89 | |
XM_006230741.1 | XP_006230803.1 | 2 - 3 | 369 | 89 | ||
GenBank mRNA | BC127524.1 | - | 2 - 3 | 327 | 7114598 | 89 |
FQ231535.1 | - | 2 - 3 | 192 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |