Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032614.2 | NP_116003.2 | 5 - 6 | 665 | 85 | |
XM_006230497.2 | XP_006230559.1 | 5 - 6 | 1078 | 85 | ||
GenBank mRNA | AF118651.1 | - | 5 - 6 | 499 | 85 | |
BC086381.1 | - | 5 - 6 | 665 | 7301390 | 85 | |
FQ216152.1 | - | 2 - 3 | 315 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |