Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012222.1 | NP_001012222.1 | 3 - 4 | 673 | 75 | |
XM_006241166.2 | XP_006241228.1 | 3 - 4 | 545 | 75 | ||
XM_006241167.2 | XP_006241229.1 | 3 - 4 | 732 | 75 | ||
XM_006241168.2 | XP_006241230.1 | 3 - 4 | 744 | 75 | ||
XM_008765259.1 | XP_008763481.1 | 3 - 4 | 605 | 75 | ||
GenBank mRNA | BC079382.1 | - | 3 - 4 | 673 | 7111095 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |