Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030868.2 | NP_110495.1 | 4 - 5 | 789 | 77 | |
GenBank mRNA | AF171936.1 | - | 4 - 5 | 776 | 77 | |
BC072548.1 | - | 4 - 5 | 789 | 7105981 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |