Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109976.2 | NP_001103446.1 | 1 - 2 | 52 | 75 | |
GenBank mRNA | CB802111.1 | - | 1 - 2 | 110 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |