Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170597.2 | NP_001164068.2 | 7 - 8 | 1949 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |