Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130507.1 | NP_001123979.1 | 2 - 3 | 229 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |