Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106298.1 | NP_001099768.1 | 4 - 5 | 697 | 134 | |
XM_006230256.2 | XP_006230318.1 | 4 - 5 | 593 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |