Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012785.3 | NP_036917.2 | 1 - 2 | 133 | 103 | |
GenBank mRNA | FQ225865.1 | - | 133 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |