Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031834.1 | NP_114022.1 | 4 - 5 | 434 | 110 | |
GenBank mRNA | AF394783.1 | - | 3 - 4 | 357 | 110 | |
FQ209630.1 | - | 4 - 5 | 443 | 110 | ||
FQ210886.1 | - | 4 - 5 | 439 | 110 | ||
FQ218459.1 | - | 4 - 5 | 439 | 110 | ||
FQ218506.1 | - | 4 - 5 | 448 | 110 | ||
FQ218658.1 | - | 4 - 5 | 438 | 110 | ||
FQ219178.1 | - | 4 - 5 | 439 | 110 | ||
FQ219521.1 | - | 4 - 5 | 439 | 110 | ||
FQ219542.1 | - | 4 - 5 | 439 | 110 | ||
FQ219948.1 | - | 4 - 5 | 439 | 110 | ||
L19998.1 | - | 4 - 5 | 434 | 110 | ||
X52883.1 | - | 4 - 5 | 422 | 110 | ||
X68640.1 | - | 3 - 4 | 339 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |