Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729998.4 | XP_002730044.2 | 6 - 7 | 793 | 77 | |
XM_006244572.2 | XP_006244634.1 | 6 - 7 | 796 | 77 | ||
XM_006244573.2 | XP_006244635.1 | 7 - 8 | 658 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |