Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007643.1 | NP_001007644.1 | 6 - 7 | 865 | 94 | |
GenBank mRNA | BC083678.1 | - | 6 - 7 | 865 | 7135830 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |