Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106485.3 | NP_001099955.2 | 1 - 2 | 287 | 79 | |
XM_006234400.2 | XP_006234462.1 | 1 - 2 | 493 | 79 | ||
GenBank mRNA | FQ229205.1 | - | 1 - 2 | 271 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |