Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057192.2 | NP_476540.2 | 2 - 3 | 354 | 69 | |
GenBank mRNA | AJ303456.1 | - | 1 - 2 | 187 | 69 | |
BC072475.1 | - | 2 - 3 | 354 | 7100751 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |