Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127525.1 | NP_001120997.1 | 1 - 2 | 109 | 60 | |
GenBank mRNA | BC161895.1 | - | 1 - 2 | 109 | 7304509 | 60 |
FQ213771.1 | - | 1 - 2 | 97 | 60 | ||
FQ220741.1 | - | 1 - 2 | 93 | 60 | ||
FQ225130.1 | - | 1 - 2 | 101 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |