Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240456.1 | XP_006240518.1 | 1 - 2 | 108 | 98 | |
GenBank mRNA | D00675.1 | - | 1 - 2 | 54 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |