Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764109.1 | XP_008762331.1 | 2 - 3 | 399 | 63 | |
XM_008764110.1 | XP_008762332.1 | 2 - 3 | 399 | 63 | ||
XM_008764111.1 | XP_008762333.1 | 2 - 3 | 124 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |