Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 6 - 7 | 2044 | 65 | |
XM_008763989.1 | XP_008762211.1 | 7 - 8 | 3346 | 65 | ||
GenBank mRNA | AY530298.1 | - | 5 - 6 | 1907 | 65 | |
AY530299.1 | - | 5 - 6 | 1907 | 65 | ||
U67139.1 | - | 6 - 7 | 2011 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |