Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145671.1 | NP_663704.1 | 1 - 2 | 168 | 136 | |
XM_006239521.2 | XP_006239583.1 | 1 - 2 | 195 | 136 | ||
XM_008764336.1 | XP_008762558.1 | 1 - 2 | 195 | 136 | ||
GenBank mRNA | AF505655.1 | - | 1 - 2 | 168 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |