Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021582.1 | NP_067593.1 | 4 - 5 | 419 | 62 | |
GenBank mRNA | BC079180.1 | - | 4 - 5 | 419 | 7130711 | 62 |
FQ228766.1 | - | 1 - 2 | 48 | 62 | ||
X98490.1 | - | 3 - 4 | 325 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |