Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007606.1 | NP_001007607.1 | 4 - 5 | 508 | 65 | |
XM_006233136.2 | XP_006233198.1 | 4 - 5 | 543 | 65 | ||
GenBank mRNA | BC061765.1 | - | 4 - 5 | 508 | 5620893 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |