Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126120.1 | NP_001119592.1 | 2 - 3 | 135 | 59 | |
GenBank mRNA | BC083724.1 | - | 2 - 3 | 135 | 59 | |
FQ212950.1 | - | 2 - 3 | 191 | 59 | ||
FQ213560.1 | - | 2 - 3 | 180 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |