Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044302.1 | NP_001037767.1 | 2 - 3 | 91 | 63 | |
XM_006233176.2 | XP_006233238.1 | 2 - 3 | 301 | 63 | ||
XM_006233177.2 | XP_006233239.1 | 2 - 3 | 132 | 63 | ||
XM_006233178.2 | XP_006233240.1 | 2 - 3 | 301 | 63 | ||
XM_006233179.2 | XP_006233241.1 | 2 - 3 | 132 | 63 | ||
GenBank mRNA | BC105874.1 | - | 2 - 3 | 91 | 7131142 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |