Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232003.1 | XP_006232065.1 | 3 - 4 | 1327 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |