Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107726.1 | NP_001101196.1 | 1 - 2 | 126 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |