Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012083.1 | NP_001012083.1 | 3 - 4 | 460 | 69 | |
XM_006233234.2 | XP_006233296.1 | 3 - 4 | 2055 | 69 | ||
XM_006233235.2 | XP_006233297.1 | 3 - 4 | 2056 | 69 | ||
GenBank mRNA | BC082033.1 | - | 3 - 4 | 460 | 7130739 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |