Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031327.2 | NP_112617.2 | 3 - 4 | 805 | 65 | |
GenBank mRNA | AB015877.1 | - | 3 - 4 | 670 | 65 | |
AF218568.1 | - | 3 - 4 | 805 | 65 | ||
BC081689.1 | - | 3 - 4 | 785 | 7122117 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |