Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729925.3 | XP_002729971.1 | 2 - 3 | 728 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |