Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271227.1 | NP_001258156.1 | 6 - 7 | 1277 | 80 | |
XM_006256721.2 | XP_006256783.1 | 6 - 7 | 1922 | 80 | ||
XM_006256722.1 | XP_006256784.1 | 5 - 6 | 1167 | 80 | ||
XM_006256723.2 | XP_006256785.1 | 6 - 7 | 1243 | 80 | ||
XM_006256725.2 | XP_006256787.1 | 6 - 7 | 936 | 80 | ||
XM_006256726.2 | XP_006256788.1 | 6 - 7 | 886 | 80 | ||
XM_006256727.2 | XP_006256789.1 | 6 - 7 | 1286 | 80 | ||
XM_006256728.2 | XP_006256790.1 | 6 - 7 | 955 | 80 | ||
XM_006256729.2 | XP_006256791.1 | 6 - 7 | 906 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |