Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033070.1 | NP_001028242.1 | 5 - 6 | 455 | 138 | |
XM_006240781.2 | XP_006240843.1 | 5 - 6 | 484 | 138 | ||
XM_008765031.1 | XP_008763253.1 | 5 - 6 | 479 | 138 | ||
GenBank mRNA | BC100070.1 | - | 5 - 6 | 455 | 7385206 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |