Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031086.2 | NP_112348.2 | 5 - 6 | 549 | 102 | |
XM_006240719.2 | XP_006240781.1 | 3 - 4 | 835 | 102 | ||
XM_008765045.1 | XP_008763267.1 | 3 - 4 | 281 | 102 | ||
GenBank mRNA | DY571157.1 | - | 4 - 5 | 370 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |