Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108812.1 | NP_001102282.1 | 6 - 7 | 661 | 123 | |
XM_006244334.2 | XP_006244396.2 | 7 - 8 | 951 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |