Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007711.1 | NP_001007712.1 | 6 - 7 | 812 | 108 | |
XM_006244321.2 | XP_006244383.1 | 7 - 8 | 935 | 108 | ||
GenBank mRNA | BC079136.1 | - | 6 - 7 | 812 | 7127699 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |