Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012558.3 | NP_036690.2 | 3 - 4 | 649 | 71 | |
GenBank mRNA | BC078894.1 | - | 3 - 4 | 631 | 7114766 | 71 |
BC078895.1 | - | 3 - 4 | 649 | 7134943 | 71 | |
J04112.1 | - | 3 - 4 | 445 | 71 | ||
M86240.1 | - | 3 - 4 | 535 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |