Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105994.1 | NP_001099464.1 | 2 - 3 | 273 | 88 | |
XM_006250541.1 | XP_006250603.1 | 2 - 3 | 326 | 88 | ||
XM_008769927.1 | XP_008768149.1 | 2 - 3 | 1185 | 88 | ||
GenBank mRNA | FQ212126.1 | - | 2 - 3 | 297 | 88 | |
FQ213206.1 | - | 2 - 3 | 199 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |