Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172084.1 | NP_001165555.1 | 4 - 5 | 573 | 103 | |
XM_008769938.1 | XP_008768160.1 | 2 - 3 | 211 | 103 | ||
GenBank mRNA | BC083854.1 | - | 4 - 5 | 509 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |