Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153624.2 | NP_705888.2 | 20 - 21 | 3140 | 69 | |
XM_006254403.2 | XP_006254465.1 | 20 - 21 | 3196 | 69 | ||
GenBank mRNA | BC166779.1 | - | 2 - 3 | 178 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |