Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014203.1 | NP_001014225.1 | 3 - 4 | 604 | 124 | |
XM_006257362.2 | XP_006257424.1 | 3 - 4 | 670 | 124 | ||
GenBank mRNA | BC083683.1 | - | 3 - 4 | 604 | 7135919 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |