Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252181.2 | XP_006252243.1 | 4 - 5 | 1209 | 133 | |
GenBank mRNA | BC158553.1 | - | 4 - 5 | 405 | 9088212 | 133 |
FQ232228.1 | - | 3 - 4 | 194 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |