Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079937.1 | NP_001073406.1 | 5 - 6 | 710 | 151 | |
XM_006252102.1 | XP_006252164.1 | 5 - 6 | 591 | 151 | ||
XM_006252103.2 | XP_006252165.1 | 5 - 6 | 569 | 151 | ||
XM_006252104.2 | XP_006252166.1 | 5 - 6 | 580 | 151 | ||
XM_006252105.2 | XP_006252167.1 | 5 - 6 | 849 | 151 | ||
XM_006252106.2 | XP_006252168.1 | 5 - 6 | 789 | 151 | ||
GenBank mRNA | BC129099.1 | - | 5 - 6 | 710 | 8375047 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |