Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270855.1 | NP_001257784.1 | 2 - 3 | 190 | 67 | |
NM_001270856.1 | NP_001257785.1 | 2 - 3 | 190 | 67 | ||
NM_001270857.1 | NP_001257786.1 | 2 - 3 | 190 | 67 | ||
NM_001270858.1 | NP_001257787.1 | 2 - 3 | 190 | 67 | ||
NM_001270859.1 | NP_001257788.1 | 2 - 3 | 121 | 67 | ||
NM_019176.2 | NP_062049.1 | 2 - 3 | 190 | 67 | ||
XM_006252149.2 | XP_006252211.1 | 3 - 4 | 344 | 67 | ||
XM_006252151.2 | XP_006252213.1 | 2 - 3 | 1253 | 67 | ||
XM_006252156.2 | XP_006252218.1 | 2 - 3 | 1249 | 67 | ||
XM_008770778.1 | XP_008769000.1 | 2 - 3 | 197 | 67 | ||
GenBank mRNA | AF026528.1 | - | 2 - 3 | 131 | 67 | |
AF026529.1 | - | 2 - 3 | 131 | 67 | ||
AF026530.1 | - | 2 - 3 | 131 | 67 | ||
BC092646.1 | - | 2 - 3 | 127 | 7319130 | 67 | |
CB580507.1 | - | 2 - 3 | 121 | 67 | ||
FQ212976.1 | - | 2 - 3 | 141 | 67 | ||
FQ213134.1 | - | 2 - 3 | 191 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |