Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107791.1 | NP_001101261.1 | 5 - 6 | 356 | 74 | |
XM_006235317.1 | XP_006235379.1 | 14 - 15 | 1316 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |