Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100477.1 | NP_001093947.1 | 9 - 10 | 1274 | 144 | |
GenBank mRNA | D50580.1 | - | 9 - 10 | 1266 | 144 | |
FQ218265.1 | - | 9 - 10 | 1314 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |