Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772340.1 | XP_008770562.1 | 3 - 4 | 528 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |