Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271091.1 | NP_001258020.1 | 8 - 9 | 720 | 61 | |
NM_001271092.1 | NP_001258021.1 | 8 - 9 | 720 | 61 | ||
NM_001271093.1 | NP_001258022.1 | 8 - 9 | 667 | 61 | ||
NM_001271094.1 | NP_001258023.1 | 8 - 9 | 667 | 61 | ||
NM_001271095.1 | NP_001258024.1 | 6 - 7 | 585 | 61 | ||
NM_031967.2 | NP_114173.2 | 6 - 7 | 585 | 61 | ||
XM_006255123.2 | XP_006255185.1 | 8 - 9 | 662 | 61 | ||
GenBank mRNA | AF045564.1 | - | 6 - 7 | 517 | 61 | |
AY217029.1 | - | 6 - 7 | 556 | 61 | ||
AY217030.1 | - | 8 - 9 | 667 | 61 | ||
AY217031.1 | - | 8 - 9 | 667 | 61 | ||
AY217032.1 | - | 8 - 9 | 715 | 61 | ||
AY217033.1 | - | 8 - 9 | 715 | 61 | ||
AY255791.1 | - | 6 - 7 | 556 | 61 | ||
CB585874.1 | - | 2 - 3 | 142 | 61 | ||
FM054107.1 | - | 6 - 7 | 538 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |