Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013888.1 | NP_001013910.1 | 1 - 2 | 165 | 93 | |
XM_006255073.2 | XP_006255135.1 | 1 - 2 | 181 | 93 | ||
GenBank mRNA | BC079217.1 | - | 1 - 2 | 165 | 7131896 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |