Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012056.1 | NP_001012056.1 | 11 - 12 | 1540 | 78 | |
GenBank mRNA | AF479659.1 | - | 11 - 12 | 1540 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |